Learn More
Electron Microscopy Sciences Aurion ImmunoGold Goat-anti-Rabbit IgG (H&L), UltraSmall 1.5 ML
SDP
Supplier:  Electron Microscopy Sciences 25100
Encompass_Preferred
Ultra small conjugates have far less influence on the adsorbed antibodies or detecting molecules, thus behave as though they are uncoupled. Used with R-Gent SE-LM and SE-EM silver enhancement reagents.
Particle size: UltraSmall
Catalog No. 50-263-02