Learn More
Electron Microscopy Sciences Aurion ImmunoGold Goat-anti-Rat IgG (H&L), UltraSmall 0.6 ML
SDP
Supplier:  Electron Microscopy Sciences 25181
Encompass_Preferred
Ultra small conjugates have far less influence on the adsorbed antibodies or detecting molecules, thus behave as though they are uncoupled. Used with R-Gent SE-LM and SE-EM silver enhancement reagents they are the best choice for any application.
Particle size: UltraSmall
Catalog No. 50-263-18