Learn More
Innovative Research Inc Goat RBCs Packed 10 15ml
Catalog No. 502744368
Encompass_Preferred
Goat Red Blood Cells Packed 10% 15ml - Goat Red Blood Cells from Innovative Research are manufactured using whole blood collections from goats. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 10% suspension. Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4368 Supplier Innovative Research Inc Supplier No. IGTRBC10P15ML
Goat Red Blood Cells Packed 10% 15ml - Goat Red Blood Cells from Innovative Research are manufactured using whole blood collections from goats. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 10% suspension. Red blood cells are perishable and are collected and processed upon receipt of order.