Learn More
Innovative Research Inc Goat RBCs Pckd 100 15ml
Catalog No. 502744369
Encompass_Preferred
Goat Red Blood Cells Packed 100% 15ml - Goat Red Blood Cells from Innovative Research are manufactured using whole blood collections from goats. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 100% suspension. Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4369 Supplier Innovative Research Inc Supplier No. IGTRBC100P15ML
Goat Red Blood Cells Packed 100% 15ml - Goat Red Blood Cells from Innovative Research are manufactured using whole blood collections from goats. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 100% suspension. Red blood cells are perishable and are collected and processed upon receipt of order.