Learn More
American Research Products Inc Goat Serum amyloid A SAA ELI
Catalog No. 5000330025
Encompass
Goat Serum amyloid A (SAA) ELISA Kit from CUSABIO Detection Range 100 ng/ml-8000 ng/ml Sensitivity 125 ng/ml Sample serum plasma Method Competitive
Catalog No. 50-003-30025 Supplier American Research Products Inc Supplier No. CSBE13212G
Goat Serum amyloid A (SAA) ELISA Kit from CUSABIO Detection Range 100 ng/ml-8000 ng/ml Sensitivity 125 ng/ml Sample serum plasma Method Competitive