Learn More
MyBioSource Goat TMPRSS2 Polyclonal Antibody, 0.1 mg
SDP
Supplier:  MyBioSource MBS421855
Encompass_Preferred
Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide. Supplied at 0.5 mg/ml in Tris saline, 0. 02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Catalog No. NC1797466