Learn More
American Research Products Inc Goat Tumor necrosis factor a
Catalog No. 5000328742
Encompass
Goat Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28742 Supplier American Research Products Inc Supplier No. CSBE09811G
Goat Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich