Learn More
Cayman Chemical GoatFAb2ntIMouseIgGF100ug
SDP
Catalog No. 502968224
Encompass_Preferred
Goat F(Ab')2 anti-Mouse IgG, Fc-specific conjugated to SureLight™ APC Antibody: Goat F(Ab')2 polyclonal anti-Mouse IgG, Fc-specifc Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry and Fluorescence-activated cell sorting
Catalog No. 50-296-8224 Supplier Cayman Chemical Supplier No. 16590100 UG
Goat F(Ab')2 anti-Mouse IgG, Fc-specific conjugated to SureLight™ APC Antibody: Goat F(Ab')2 polyclonal anti-Mouse IgG, Fc-specifc Dye: SureLight™ Allophycocyanin (APC) Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry and Fluorescence-activated cell sorting