Learn More
Cayman Chemical GoatntIMouseIgG2bSureLIg1ea
SDP
Catalog No. 502955508
Encompass_Preferred
Goat anti-Mouse IgG2b conjugated to SureLight APC, 100μg Antibody: Goat polyclonal anti-Mouse IgG2b Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry/Fluorescence-activated cell sorting
Catalog No. 50-295-5508 Supplier Cayman Chemical Supplier No. 133971 EA
Goat anti-Mouse IgG2b conjugated to SureLight APC, 100μg Antibody: Goat polyclonal anti-Mouse IgG2b Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry/Fluorescence-activated cell sorting