Learn More
Cayman Chemical GoatntIMouseIgGSureLIght1ea
SDP
Catalog No. 502955779
Encompass_Preferred
Goat anti-mouse IgG conjugated to SureLight P3 Antibody: Goat polyclonal anti-Mouse IgG Dye: SureLight P3 Excitation max. λ: 614 nm Emission max. λ: 662 nm Isotype: IgG Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays
Catalog No. 50-295-5779 Supplier Cayman Chemical Supplier No. 134441 EA
Goat anti-mouse IgG conjugated to SureLight P3 Antibody: Goat polyclonal anti-Mouse IgG Dye: SureLight P3 Excitation max. λ: 614 nm Emission max. λ: 662 nm Isotype: IgG Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays