Learn More
MSC Steiner Welding Gloves, Goatskin Leather, Large, MIG, 12 inch
Catalog No. 502835143
Encompass
These welding gloves are designed for MIG welding applications, offering a large size for comfortable fit and a 12 inch overall length. Constructed with goatskin leather and featuring a gauntlet cuff, they provide flame protection and a reinforced palm for enhanced durability and safety.
  • Welding glove type
  • MIG welding application
  • Large glove size
  • Goatskin leather material
  • Lined with cotton
  • Gauntlet cuff style
  • 12 inch overall length
  • Wing thumb type
  • Seamless lining
  • Premium grain leather
  • Flame protection
  • Heavy fabric weight
  • Reinforced palm
Catalog No. 50-283-5143 Supplier MSC Supplier No. 40742926
These welding gloves are designed for MIG welding applications, offering a large size for comfortable fit and a 12 inch overall length. Constructed with goatskin leather and featuring a gauntlet cuff, they provide flame protection and a reinforced palm for enhanced durability and safety.
  • Welding glove type
  • MIG welding application
  • Large glove size
  • Goatskin leather material
  • Lined with cotton
  • Gauntlet cuff style
  • 12 inch overall length
  • Wing thumb type
  • Seamless lining
  • Premium grain leather
  • Flame protection
  • Heavy fabric weight
  • Reinforced palm