Learn More
MSC ProTeam Backpack Vacuum, GoFit 6 qt., Remediation Kit, ProLevel Filtration
Catalog No. 502835356
Encompass
The ProTeam GoFit Series backpack vacuum offers commercial cleaning performance with enhanced user comfort. Featuring a FlexFit harness and ProLevel Filtration, it efficiently cleans various surfaces. This model includes a specialized remediation tool kit.
  • FlexFit articulating harness for increased comfort.
  • Advanced filtration system for capturing harmful particles.
  • Efficiently cleans carpets and hard surfaces.
  • Includes a specialized remediation tool kit.
  • 6 quart capacity.
  • 153 CFM airflow.
  • ProLevel Filtration with HEPA Media.
  • UL Listed and CRI Seal of Approval.
Catalog No. 50-283-5356 Supplier MSC Supplier No. 97101448
The ProTeam GoFit Series backpack vacuum offers commercial cleaning performance with enhanced user comfort. Featuring a FlexFit harness and ProLevel Filtration, it efficiently cleans various surfaces. This model includes a specialized remediation tool kit.
  • FlexFit articulating harness for increased comfort.
  • Advanced filtration system for capturing harmful particles.
  • Efficiently cleans carpets and hard surfaces.
  • Includes a specialized remediation tool kit.
  • 6 quart capacity.
  • 153 CFM airflow.
  • ProLevel Filtration with HEPA Media.
  • UL Listed and CRI Seal of Approval.