Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ProTeam Backpack Vacuum, 6 qt, Cordless, 4Ah, HEPA Filtration

Catalog No. 502835058
Encompass

Go wherever tasks take you with the ProTeam GoFit Cordless backpack vacuum, 6 qt. This cordless model allows safe maneuverability around obstacles.

  • Cordless design: Cleans 30% faster than standard backpack vacuums and 5 times faster than uprights.
  • Enhanced mobility: Navigate spaces freely without cord restrictions.
  • Extended runtimes: Up to 51 minutes on low power with the 4Ah battery.
  • Lightweight: Weighs just 13.5 pounds with battery for comfortable use.
  • Dual power modes: High/low switch for detailed cleaning or high-traffic areas.
  • Fast charging: Battery fully recharges in approximately 61 minutes.
  • Custom comfort: Quick-adjust 4-step harness for a personalized fit.
  • Superior filtration: ProLevel Filtration System captures soil and harmful particles.
  • Safer cleaning: Cordless operation eliminates tripping hazards.

Catalog No. 50-283-5058 Supplier MSC Supplier No. 97096762
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Go wherever tasks take you with the ProTeam GoFit Cordless backpack vacuum, 6 qt. This cordless model allows safe maneuverability around obstacles.

  • Cordless design: Cleans 30% faster than standard backpack vacuums and 5 times faster than uprights.
  • Enhanced mobility: Navigate spaces freely without cord restrictions.
  • Extended runtimes: Up to 51 minutes on low power with the 4Ah battery.
  • Lightweight: Weighs just 13.5 pounds with battery for comfortable use.
  • Dual power modes: High/low switch for detailed cleaning or high-traffic areas.
  • Fast charging: Battery fully recharges in approximately 61 minutes.
  • Custom comfort: Quick-adjust 4-step harness for a personalized fit.
  • Superior filtration: ProLevel Filtration System captures soil and harmful particles.
  • Safer cleaning: Cordless operation eliminates tripping hazards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.