Learn More
MSC ProTeam Backpack Vacuum, 3 qt. Capacity, 120 V, HEPA Filtration
Catalog No. 502834832
Encompass
This backpack vacuum offers powerful and efficient cleaning, featuring a a 1,330 W motor with 153 CFM airflow and 91 in static lift, operating quietly at 68 dB. Designed for comfort and maneuverability in tight spaces, it includes a multi-surface floor tool and a 2-piece aluminum wand. Its ProLevel HEPA filtration system captures 99.79% of particulates, reducing allergens. Easy filter changes are facilitated by a hinged lid with a quick-release latch. The unit requires a 120 V connection.
  • Ergonomic 4-step FlexFit articulating harness for comfortable wear.
  • Low-profile design suited for under furniture and tight spaces.
  • Includes professional-grade hose and a 50 ft. power cord.
  • Comes with two micro filter bags, a micro cloth filter, a dome filter, and an exhaust filter.
Catalog No. 50-283-4832 Supplier MSC Supplier No. 97104400
This backpack vacuum offers powerful and efficient cleaning, featuring a a 1,330 W motor with 153 CFM airflow and 91 in static lift, operating quietly at 68 dB. Designed for comfort and maneuverability in tight spaces, it includes a multi-surface floor tool and a 2-piece aluminum wand. Its ProLevel HEPA filtration system captures 99.79% of particulates, reducing allergens. Easy filter changes are facilitated by a hinged lid with a quick-release latch. The unit requires a 120 V connection.
  • Ergonomic 4-step FlexFit articulating harness for comfortable wear.
  • Low-profile design suited for under furniture and tight spaces.
  • Includes professional-grade hose and a 50 ft. power cord.
  • Comes with two micro filter bags, a micro cloth filter, a dome filter, and an exhaust filter.