Learn More
MSC ProTeam Backpack Vacuum, GoFit Series, HEPA Filtration, 6 qt
Catalog No. 502817239
Encompass
This portable and backpack vacuum cleaner is designed for efficient cleaning, operating on 120V with 11.70 Amps. It comes with a comprehensive kit of tools for various cleaning tasks, ensuring versatility and ease of use.
  • Electric power source
  • HEPA filtration for improved air quality
  • Disposable bag collection type
  • Maximum air flow of 153 CFM
  • Tank capacity of 6 qt
  • Low sound level of 67.9 db(A)
  • Includes 50 ft power cord
  • Comes with a 42 to 57 in telescoping wand and various floor tools
Catalog No. 50-281-7239 Supplier MSC Supplier No. 97097349
This portable and backpack vacuum cleaner is designed for efficient cleaning, operating on 120V with 11.70 Amps. It comes with a comprehensive kit of tools for various cleaning tasks, ensuring versatility and ease of use.
  • Electric power source
  • HEPA filtration for improved air quality
  • Disposable bag collection type
  • Maximum air flow of 153 CFM
  • Tank capacity of 6 qt
  • Low sound level of 67.9 db(A)
  • Includes 50 ft power cord
  • Comes with a 42 to 57 in telescoping wand and various floor tools