Learn More
MSC ProTeam GoFit Cordless Battery, 4Ah, Lithium Ion
Catalog No. 502837411
Encompass
Experience uninterrupted cleaning sessions with this 4Ah Lithium-Ion battery. When used with a compatible cordless unit, its high/low power switch enables approximately 51 minutes of runtime on low setting for routine or detail cleaning.
  • Cordless efficiency for faster cleaning
  • Provides mobility without cord restrictions
  • High/low power switch to conserve runtime
  • Easy-to-read battery fuel gauge displays approximate remaining runtime
  • Minimum of 1000 charge cycles for extended life
  • Fast charge; reaches full charge in approximately 61 minutes
  • Long runtime with approximately 51 minutes on low setting and 39 minutes on high setting
  • Long-lasting Lithium-Ion battery with a control circuit
  • Seamless battery swap for quick replacement
  • Compatible with GoFit Cordless vacuums
  • 2-year warranty
Catalog No. 50-283-7411 Supplier MSC Supplier No. 40608200
Experience uninterrupted cleaning sessions with this 4Ah Lithium-Ion battery. When used with a compatible cordless unit, its high/low power switch enables approximately 51 minutes of runtime on low setting for routine or detail cleaning.
  • Cordless efficiency for faster cleaning
  • Provides mobility without cord restrictions
  • High/low power switch to conserve runtime
  • Easy-to-read battery fuel gauge displays approximate remaining runtime
  • Minimum of 1000 charge cycles for extended life
  • Fast charge; reaches full charge in approximately 61 minutes
  • Long runtime with approximately 51 minutes on low setting and 39 minutes on high setting
  • Long-lasting Lithium-Ion battery with a control circuit
  • Seamless battery swap for quick replacement
  • Compatible with GoFit Cordless vacuums
  • 2-year warranty