Learn More
MSC ProTeam Backpack Vacuum, 6 qt, HEPA Filtration, 120V
Catalog No. 502835312
Encompass
This portable and backpack vacuum cleaner operates on electric power with 120V and 11.70 amps. It features HEPA filtration and a 6 qt collection capacity, utilizing a disposable bag for vacuum collection. With a maximum air flow of 153 CFM and a 50 ft power cord, this vacuum is designed for comfortable use and efficient cleaning in various environments.
  • Reimagined 4-step FlexFit harness provides comfort and breathability.
  • Quick-adjust harness designed for a wide range of users.
  • Advanced filtration system captures soil and harmful particles.
  • Efficiently cleans carpets and hard surfaces, reducing operator fatigue.
  • Reduced profile triangular design maximizes comfort and productivity.
  • Improved cord management with an intuitive switchbox.
  • Includes a versatile telescoping wand with a carpet floor tool.
  • 3-year warranty on motor, labor, and parts.
Catalog No. 50-283-5312 Supplier MSC Supplier No. 97101794
This portable and backpack vacuum cleaner operates on electric power with 120V and 11.70 amps. It features HEPA filtration and a 6 qt collection capacity, utilizing a disposable bag for vacuum collection. With a maximum air flow of 153 CFM and a 50 ft power cord, this vacuum is designed for comfortable use and efficient cleaning in various environments.
  • Reimagined 4-step FlexFit harness provides comfort and breathability.
  • Quick-adjust harness designed for a wide range of users.
  • Advanced filtration system captures soil and harmful particles.
  • Efficiently cleans carpets and hard surfaces, reducing operator fatigue.
  • Reduced profile triangular design maximizes comfort and productivity.
  • Improved cord management with an intuitive switchbox.
  • Includes a versatile telescoping wand with a carpet floor tool.
  • 3-year warranty on motor, labor, and parts.