Learn More
MSC ProTeam Backpack Vacuum, GoFit 3 Cordless, 3 qt, HEPA Filtration
Catalog No. 502836996
Encompass
The ProTeam GoFit 3 Cordless Backpack Vacuum is designed for commercial cleaning, offering enhanced mobility and efficiency. It features a battery power source and HEPA filtration.
  • Cleans 30% faster than standard backpack vacuums and 5 times faster than uprights.
  • Offers enhanced mobility without cord restrictions.
  • Provides extended runtime: approximately 106 minutes on low, 83 minutes on high.
  • Lightweight design, only 15.7 pounds with battery.
  • High/low power switch for detailed or high-traffic area cleaning.
  • Rapid battery recharge in about 101 minutes.
  • Features an interchangeable, tool-free 4-step harness for comfort.
  • Advanced filtration system captures soil and harmful particles.
  • Eliminates cord tripping hazards for a safer cleaning environment.
  • Multi-surface cleaning capability.
Catalog No. 50-283-6996 Supplier MSC Supplier No. 40614166
The ProTeam GoFit 3 Cordless Backpack Vacuum is designed for commercial cleaning, offering enhanced mobility and efficiency. It features a battery power source and HEPA filtration.
  • Cleans 30% faster than standard backpack vacuums and 5 times faster than uprights.
  • Offers enhanced mobility without cord restrictions.
  • Provides extended runtime: approximately 106 minutes on low, 83 minutes on high.
  • Lightweight design, only 15.7 pounds with battery.
  • High/low power switch for detailed or high-traffic area cleaning.
  • Rapid battery recharge in about 101 minutes.
  • Features an interchangeable, tool-free 4-step harness for comfort.
  • Advanced filtration system captures soil and harmful particles.
  • Eliminates cord tripping hazards for a safer cleaning environment.
  • Multi-surface cleaning capability.