Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ GoGel™ Agarose Gel Pack (10 Gel Packs)
SDPSCI_ED

Catalog No. S05964 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
10 Pack, 45 mL each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05964 10 Pack, 45 mL each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05964 Supplier EDVOTEK™ Supplier No. 603
Add to Cart
Add to Cart

Introducing the Edvotek GoGel™ Agarose Gel Pack - the perfect solution for the busy biotechnology teacher. Our innovative, single use gel packs are designed for ease and convenience, containing meltable 0.8% agarose gel, mixed with TAE buffer, and SYBR™ Safe DNA Stain.

  • To prepare the gel, just tear open the pack, squeeze the gel into a microwave safe container, and heat until the solid melts. Allow to cool to 60°C before pouring into the gel casting system. With GoGel™, you will have a hassle free gel preparation experience!
  • Each pack contains 45 mL of 0.8% agarose gel in 1X TAE with SYBR™ Safe fluorescent DNA Stain
  • 10 Gel Packs per order
  • 50x TAE concentrate sold separately
  • Storage: Refrigerator

Specifications

Buffer 1X TAE
Color Clear, Colorless
Format Melt and Pour
Gel Percentage 0.8%
Gel Size Mini
Label or Dye SYBR Safe
Separation of DNA
Separation Type Electrophoresis
Wells 1-well
For Use With (Application) Agarose
Quantity 10 Pack, 45 mL each
Shipping Condition Ambient
Sterility Sterile
Storage Requirements 4°C
Product Type GoGel™ Agarose Gel Pack (10 Gel Packs)
For Use With (Equipment) Mini Gel Tank
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.