Learn More
Med Vet International ProtectIVe Goggles 3M Clear Tint Polycarbonate Lens Elastic Strap One Size Fits Most, 40660-00000-10, 10/Box
Catalog No. 502830214
Encompass_Preferred
Dependable, fully compliant splash protection Tough Duralite polycarbonate lens absorbs 99.9% of UV rays. Indirect venting circulates air to lens while keeping liquids out Large lens opening fits over most prescription or safety glasses
Catalog No. 50-283-0214 Supplier Med Vet International Supplier No. 697106BX
Dependable, fully compliant splash protection Tough Duralite polycarbonate lens absorbs 99.9% of UV rays. Indirect venting circulates air to lens while keeping liquids out Large lens opening fits over most prescription or safety glasses