Learn More
3M™ GoggleGear™ 500-Series
Catalog No. 19040627 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog, UV-treated
Catalog No. Lens Coating
19-040-627 Anti-fog, UV-treated
1 options
This item is not returnable. View return policy
Catalog No. 19-040-627 Supplier 3M™ Supplier No. 7100168468
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ GoggleGear™ 500-Series helps keep lenses clear in rugged, dusty, steamy and wet environments by providing longer lasting fog resistance.
  • Workers may enjoy reliable, clearer sight longer, because the coating with its active ingredient is bonded to the lens.
  • The coating retains its effectiveness for at least 25 washings with water, allowing workers to wear their eyewear longer
  • The coating can be disinfected with diluted bleach soaking or alcohol wipes without losing its anti-fog performance.
  • Washing with soap and water is considered a best practice and can enhance coating durability.
  • The coating withstands disinfection with diluted bleach soaking or alcohol wipes without losing its anti-fog performance
  • 3M Catalog# GG502SGAF
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
Product Type Safety Goggles
Frame Color Gray, Red
Model GG502SGAF
No. per Pack 10/Pk.
Lens Material Polycarbonate
UV Protection UV400