Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ GoggleGear™ 500 Series Prescription Insert

Catalog No. 19169039 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M Goggle Gear, 500-Series Splash Goggles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-169-039 3M Goggle Gear, 500-Series Splash Goggles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-039 Supplier 3M™ Supplier No. 7100084923
  • $16.00
    $14.65 / Each

    Save $1.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $158.00
    $146.65 / Case of 10 EA

    Save $11.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Offers the option to have a prescription lens made to go with 3M GoggleGear, 500-Series Splash Goggles. Helpful for anyone who needs corrective vision to go along with eye protection.

  • Insert is coated with Scotchgard™ Anti-Fog coating which retains its effectiveness for at least 25 washings with water, allowing workers to wear their eyewear longer.
  • 3M Catalog# GG500-PI

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Product Type Prescription Insert
For Use With (Equipment) 3M Goggle Gear, 500-Series Splash Goggles
Model GG500-PI
Material Polycarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.