Description
- Scotchgard anti-fog lens keep lenses clear in rugged, dusty, steamy and wet environments providing longer lasting fog resistance than traditional anti-fog, anti-scratch coatings
- Low-profile design with an adjustable neoprene strap and indirect ventilation
- Coating can be disinfected with diluted bleach soaking or alcohol wipes without losing its anti-fog performance
- Meets the requirements of ANSI Z87.1 and CSA Z94.3
- 3M Catalog# GG501NSGAF
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | ANSI Z87.1-2003, CSA Z94.3-2007 |
| Product Type | Safety Goggles |
| Frame Color | Gray, Red |
| Model | GG501NSGAF |
| Lens Material | Polycarbonate |
| UV Protection | UV400 |