Learn More
3M™ GoggleGear™ 6000 Series Scotchgard™ Anti-Fog Coating Replacement Lens
Catalog No. 19071102 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog, UV-treated
Catalog No. Lens Coating
19-071-102 Anti-fog, UV-treated
1 options
This item is not returnable. View return policy
Catalog No. 19-071-102 Supplier 3M™ Supplier No. 7100191927
This item is not returnable. View return policy
3M™ Scotchgard™ Anti-Fog Coating helps increase eyewear performance in wet and steamy environments
  • Scotchgard™ Anti-fog coating for fog and abrasion resistance
  • Polycarbonate lens absorbs 99.9% UVA & UVB and meet the requirements of ANSI Z87.1- 2015 Standard as an Impact Rated spectacle
  • All models come with indirect venting to help reduce fogging
  • Both adjustable nylon fiber cloth strap, or neoprene strap, are available within the 3M™ GoggleGear™ 6000 Series.
  • 3M Catalog# GG6001SGAF-LENS
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Replacement lens
For Use With (Application) General Manufacturing, Grinding, Construction, Pharmaceuticals, Oil and Gas
No. per Case 30/Cs.
Model GG6001SGAF-LENS
Material Polycarbonate
Lens Material Polycarbonate
UV Protection UV400