Learn More
3M™ GoggleGear™ 6000 Series Scotchgard™ Goggles
Catalog No. 19071099 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Black
Gray
Green
Lens Tint:
Clear
Gray
Headband Material:
Neoprene
Nylon
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Headband Material
19-071-099 Black Gray Nylon
19-071-096 Black Clear Nylon
19-071-097 Green Clear Nylon
19-071-100 Gray Clear Neoprene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-071-099 Supplier 3M™ Supplier No. 7100191930
  • $539.00
    $324.65 / Case of 10
    Save $214.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Scotchgard™ Anti-Fog Coating helps to increase eyewear performance in wet and steamy environments.
  • Indirect venting allows air to circulate and helps reduce fogging
  • D3 (droplet and splash) and D4 (dust) rated
  • Scotchgard™ Anti-fog coating for fog and abrasion resistance
  • Polycarbonate lens absorbs 99.9% UVA & UVB and meet the requirements of ANSI Z87.1- 2015 Standard as an Impact Rated spectacle
  • 3M Catalog# GG6001NSGAF-BLU, GG6001SGAF-BLK, GG6001SGAF-GRN, GG6001SGAF-RED, GG6002SGAF-BLK
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
Product Type Safety Goggles
For Use With (Application) Chemical Handling, Painting, Primary Metals, General Manufacturing, Oil and Gas
Frame Color Black
Headband Material Nylon
No. per Case 10/Cs.
Model GG6002SGAF-BLK
Size Universal
Ventilation Details Indirect venting
Material Polycarbonate
Lens Material Polycarbonate
UV Protection UV400