Learn More
3M™ GoggleGear™ Safety Goggles
Catalog No. 17000689 Shop All 3M Products
This item is not returnable. View return policy
Catalog No. 17-000-689 Supplier 3M™ Supplier No. 7010386529
This item is not returnable. View return policy
Provides eye protection with indirect venting. The GoggleGear™ Safety Goggles are anti-scratch and anti-fog.
  • Indirect venting
  • Anti-scratch and Scotchgard Anti-fog coating
  • Clear polycarbonate lens
  • D3 (droplet and splash) and D4 (dust) rated
  • 3M Catalog# GG2891-SGAF
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant, UV-treated
Lens Type Splash protection
Certifications/Compliance ANSI Z87.1-2015, D3(Droplet & Splash), D4(Dust)
Lens Tint Clear
Product Type Goggles
Frame Color Gray
Headband Material Cloth
No. per Case 10/Cs.
Model GG2891-SGAF
Ventilation Details Indirect venting
Lens Material Polycarbonate
UV Protection UV400