Learn More
Benchmark Products Inc Benchmark Products A-50B-IR Goggles, Validated Sterile, Anti-Fog/A-Scratch Lens, Single-Use
SDP
Catalog No. NC9586089
Encompass
The Benchmark Products A-50B-IR Goggles are sterile, single-use, gamma-irradiated goggles designed for critical sterile environments. Featuring anti-fog and anti-scratch lenses, they ensure clear visibility and comfort while preventing cross-contamination. The low-linting adjustable elastic strap and soft polycarbonate construction provide a secure fit, making these goggles cleanroom compatible and compliant with ANSI-Z87.1 safety standards. Each pair is individually packaged for convenience and hygiene, enhancing safety in medical and laboratory settings.
This item is not returnable. View return policy
Catalog No. NC9586089 Supplier Benchmark Products Inc Supplier No. A50BIR
The Benchmark Products A-50B-IR Goggles are sterile, single-use, gamma-irradiated goggles designed for critical sterile environments. Featuring anti-fog and anti-scratch lenses, they ensure clear visibility and comfort while preventing cross-contamination. The low-linting adjustable elastic strap and soft polycarbonate construction provide a secure fit, making these goggles cleanroom compatible and compliant with ANSI-Z87.1 safety standards. Each pair is individually packaged for convenience and hygiene, enhancing safety in medical and laboratory settings.