Learn More
Hach Company Goggles, Safety, FogGard Plus
Supplier:  Hach Company 2928002
Encompass
Ply-able PVC construction means a snug, comfortable fit, even over prescription eyewear. Clear, optical-grade polycarbonate lens has a spherical design for a 160° field of vision. Choose standard lens or FogGard Plus lens, which reduces fogging in humid environments. Ultra clear lenses and blue tinted frames. Meets ANSI Z87.1 + Impact Standards.
Catalog No. 50-199-6804