Learn More
Hach Company Goggles, Safety, Uvex, Classic
Catalog No. 501996802 Shop All Hach Company Products
Encompass
Indirect ventilation system to allow free air flow through the goggle body. Large, soft body ensures an excellent fit over most prescription eyewear. Clear polycarbonate lens treated with patented Uvextreme AF coating—protects against fogging, scratching, static and dust, and 99% of harmful ultraviolet rays up to 385nm. Absorbs 99.9% of UV up to 385nm and meet ANSI Z87+ and CSA Z94.3
Catalog No. 50-199-6802 Supplier Hach Company Supplier No. 2927902
Indirect ventilation system to allow free air flow through the goggle body. Large, soft body ensures an excellent fit over most prescription eyewear. Clear polycarbonate lens treated with patented Uvextreme AF coating—protects against fogging, scratching, static and dust, and 99% of harmful ultraviolet rays up to 385nm. Absorbs 99.9% of UV up to 385nm and meet ANSI Z87+ and CSA Z94.3