Learn More
MSC Uvex Safety Goggle, V-Maxx, Anti-Fog, Clear, Direct Vent, 1 ea
Catalog No. NC2915454
Encompass
The V-Maxx Safety Goggle features sleek wrap-around styling for a 180-degree, clear field of vision. It has pivoting headband clips to adjust the strap around hard hats or hearing protection and can be worn over most prescription eyewear. This model includes direct ventilation for impact applications.
  • Sleek wrap-around styling for a 180-degree, clear field of vision
  • Pivoting headband clips to adjust strap around hard hats or hearing protection
  • Can be worn over most prescription eyewear (OTG compatible)
  • Direct ventilation for impact applications
  • Clear lens with Fog-Ban anti-fog and scratch-resistant coating
Catalog No. NC2915454 Supplier MSC Supplier No. 04498911
The V-Maxx Safety Goggle features sleek wrap-around styling for a 180-degree, clear field of vision. It has pivoting headband clips to adjust the strap around hard hats or hearing protection and can be worn over most prescription eyewear. This model includes direct ventilation for impact applications.
  • Sleek wrap-around styling for a 180-degree, clear field of vision
  • Pivoting headband clips to adjust strap around hard hats or hearing protection
  • Can be worn over most prescription eyewear (OTG compatible)
  • Direct ventilation for impact applications
  • Clear lens with Fog-Ban anti-fog and scratch-resistant coating