Learn More
LAMDA BIOTECH INC GoGreen Taq Plus Master Mix
Catalog No. NC3726336
Encompass_Preferred
A pre-formulated 2X PCR master mix containing Taq Plus DNA polymerase and integrated tracking dyes for direct gel loading capability. Designed for efficient DNA amplification with visual monitoring of reaction setup and electrophoresis.
Catalog No. NC3726336 Supplier LAMDA BIOTECH INC Supplier No. GD124
A pre-formulated 2X PCR master mix containing Taq Plus DNA polymerase and integrated tracking dyes for direct gel loading capability. Designed for efficient DNA amplification with visual monitoring of reaction setup and electrophoresis.