Learn More
Biolegend GoInVivo™ Purified anti-human CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 329727
Encompass_Preferred
GoInVivo™ Purified anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IHC, Block; Size: 1 g
Catalog No. 50-207-5897
Add to Cart
Request a Quote