Learn More
Biolegend GoInVivo™ Purified anti-mouse/rat IL-1β
Supplier:  Biolegend 503508
Encompass_Preferred
GoInVivo™ Purified anti-mouse/rat IL-1β [B122]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: ELISA Capture, ELISPOT Capture, Depletion, IHC-F, IP, Neut, WB; Size: 25 mg
Catalog No. 50-208-5597