Learn More
Biolegend GoInVivo™ Purified Armenian Hamster IgG Isotype Ctrl
Supplier:  Biolegend 400965
Encompass_Preferred
GoInVivo™ Purified Armenian Hamster IgG Isotype Ctrl [HTK888]; Isotype: Armenian Hamster IgG; Apps: FC, ICFC, ICC, IHC, IP, WB, FA; Size: 25 mg
Catalog No. 50-207-9583