Learn More
Biolegend GoInVivo™ Purified Mouse IgG1, κ Isotype Ctrl
Supplier:  Biolegend 400190
Encompass_Preferred
GoInVivo™ Purified Mouse IgG1, κ Isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC, ICFC, ICC, IHC, IP, WB, FA; Size: 500 mg
Catalog No. 50-207-9439
Add to Cart
Request a Quote