Learn More
Biolegend GoInVivo™ Purified Rat IgG2a, κ Isotype Ctrl
Supplier:  Biolegend 400560
Encompass_Preferred
GoInVivo™ Purified Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC, WB, IP, ICC, IHC, FA; Size: 500 mg
Catalog No. 50-207-9506
Add to Cart
Request a Quote