Learn More
Enterprise Technology Solutions Gojo Dispenser, Touch-free, F/REDIFOAM Soap, 2000ml Cap, Chrome
SDP
Catalog No. 5000831234
Encompass_Preferred
Touch-free Top-fill Fixture for PURELL Professional REDIFOAM Foam Soap eliminates the need to remove panels to refill soap, reducing labor by 57 percent and eliminating safety concerns. CXR Redifoam Dispensing Fixture offers fully waterproof electronics and commercial-grade construction above and below the counter with all-metal components built to handle frequent use. Patented, level-sensing design with on-spout refill indicator lights eliminate the need to manually reset. Medical-grade connections maintain an airless fluid transfer with no drips or spills. Remote foam generation ensures the system doesn't lose its prime. Choose from multiple power options, including AC adapters, hard-wire adaptation kits or batteries. Capacity is 2000 ml.
Catalog No. 50-008-31234 Supplier Enterprise Technology Solutions Supplier No. GOJ864001
Touch-free Top-fill Fixture for PURELL Professional REDIFOAM Foam Soap eliminates the need to remove panels to refill soap, reducing labor by 57 percent and eliminating safety concerns. CXR Redifoam Dispensing Fixture offers fully waterproof electronics and commercial-grade construction above and below the counter with all-metal components built to handle frequent use. Patented, level-sensing design with on-spout refill indicator lights eliminate the need to manually reset. Medical-grade connections maintain an airless fluid transfer with no drips or spills. Remote foam generation ensures the system doesn't lose its prime. Choose from multiple power options, including AC adapters, hard-wire adaptation kits or batteries. Capacity is 2000 ml.