Learn More
Enterprise Technology Solutions Gojo Foam Hand Sanitizer, f/PURELL LTX-12, 1200ml, CL
SDP
Catalog No. 5000810790
Encompass_Preferred
Foam hand-sanitizer refill is designed for PURELL LTX-12 Hands-Free Dispensers and features a scientifically advanced, patent-pending formulation that outperforms other hand sanitizers ounce for ounce. PURELL Advanced Green Certified Foam kills 99.99 percent of most common germs on hands. Green certified and USDA certified, bio-based formulation meets EcoLogo hand sanitizer standard CCD-170 for environmental leadership and proven performance as well as CDC and APIC recommendations. Nonaerosol foam formula is fragrance-free and dye-free. Hand sanitizer is clinically proven to help maintain skin health with a nourishing blend of four skin conditioners. Use with latex, nitrile and vinyl gloves, and CHG. Controlled-collapse refill (not included) holds its shape as it empties for a more attractive appearance. Sanitary-sealed refill helps prevent contamination.
Catalog No. 50-008-10790 Supplier Enterprise Technology Solutions Supplier No. GOJ190402
Foam hand-sanitizer refill is designed for PURELL LTX-12 Hands-Free Dispensers and features a scientifically advanced, patent-pending formulation that outperforms other hand sanitizers ounce for ounce. PURELL Advanced Green Certified Foam kills 99.99 percent of most common germs on hands. Green certified and USDA certified, bio-based formulation meets EcoLogo hand sanitizer standard CCD-170 for environmental leadership and proven performance as well as CDC and APIC recommendations. Nonaerosol foam formula is fragrance-free and dye-free. Hand sanitizer is clinically proven to help maintain skin health with a nourishing blend of four skin conditioners. Use with latex, nitrile and vinyl gloves, and CHG. Controlled-collapse refill (not included) holds its shape as it empties for a more attractive appearance. Sanitary-sealed refill helps prevent contamination.