Learn More
Enterprise Technology Solutions Gojo Hand Sanitizer,Gel,f/CS8 Touch-Free,VF Plus,1200ml,2/CT,CL
SDP
Catalog No. 5000811230
Encompass_Preferred
With broad-spectrum activity, PURELL VF Plus Hand Sanitizer Gel Refills are designed for PURELL CS8 Touch-free Hand Sanitizer Dispensers for professional environments. This sanitizer kills 99.99 percent of germs, including the most common foodborne, bacterial pathogens. It helps reduce germs on hands in high-traffic, high-touch areas, including restaurants and cruise ships. Quick-drying formula feels as clean as PURELL Advanced Hand Sanitizer. It meets Food Code Hand Sanitizers criteria. Hand sanitizer is fragrance-free and dye-free. It is compatible with nitrile, vinyl and latex gloves. With "Energy-on-the-refill" technology, these refills have a built-in energy source designed to last as long as the refill. AT-A-GLANCE refill design lets you monitor soap levels with just one look. Sanitary Sealed PET refill is easily recycled.
Catalog No. 50-008-11230 Supplier Enterprise Technology Solutions Supplier No. GOJ789902
With broad-spectrum activity, PURELL VF Plus Hand Sanitizer Gel Refills are designed for PURELL CS8 Touch-free Hand Sanitizer Dispensers for professional environments. This sanitizer kills 99.99 percent of germs, including the most common foodborne, bacterial pathogens. It helps reduce germs on hands in high-traffic, high-touch areas, including restaurants and cruise ships. Quick-drying formula feels as clean as PURELL Advanced Hand Sanitizer. It meets Food Code Hand Sanitizers criteria. Hand sanitizer is fragrance-free and dye-free. It is compatible with nitrile, vinyl and latex gloves. With "Energy-on-the-refill" technology, these refills have a built-in energy source designed to last as long as the refill. AT-A-GLANCE refill design lets you monitor soap levels with just one look. Sanitary Sealed PET refill is easily recycled.