Learn More
Enterprise Technology Solutions Gojo Hand Sanitizer,Gel,f/Touch-free ES6 Dispenser,1200ml,2/CT,CL
SDP
Supplier:  Enterprise Technology Solutions GOJ646102
Encompass_Preferred
E3-rated, no-rinse hand sanitizer gel is designed for use throughout food-processing facilities and fits in your PURELL ES6 Touch-free Hand Sanitizer Dispenser. It kills 99.99 percent of germs, including the most common foodborne bacterial pathogens. Listed with NSF International as an E3 for use in all departments of USDA-regulated food-processing environments, PURELL Advanced Hand Sanitizer E3 Gel is approved for hand/food contact after handwashing. It is EcoLogo Certified to Standard Name UL 2783 and a USDA Certified Biobased Product. Certified Kosher formula is hypoallergenic, dermatologist-tested, dye-free and fragrance-free. AT-A-GLANCE refill design lets you monitor soap levels with just one look. Sanitary Sealed PET refill helps prevent contamination and is easily recycled.
Catalog No. 50-008-11055