Learn More
Enterprise Technology Solutions Gojo Handwash,Foam,Antimicrobial,f/PROVON LTX-12,1200ml,2/CT,BE
SDP
Catalog No. 5000811069
Encompass_Preferred
High-performance, antimicrobial foam soap contains added moisturizers for general handwashing. Triclosan-free formulation is made with 0.3 percent chloroxylenol (PCMX), a common antimicrobial compound to help control bacteria. Foaming hand soap offers an appealing, translucent amber color with light fruity fragrance. 1200 ml. refill is designed for use with PROVON LTX-12 Dispensers. Sanitary Sealed refill helps prevent contamination. Each refill includes a fresh dispensing valve.
Catalog No. 50-008-11069 Supplier Enterprise Technology Solutions Supplier No. GOJ194402
High-performance, antimicrobial foam soap contains added moisturizers for general handwashing. Triclosan-free formulation is made with 0.3 percent chloroxylenol (PCMX), a common antimicrobial compound to help control bacteria. Foaming hand soap offers an appealing, translucent amber color with light fruity fragrance. 1200 ml. refill is designed for use with PROVON LTX-12 Dispensers. Sanitary Sealed refill helps prevent contamination. Each refill includes a fresh dispensing valve.