Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ MULTI GREEN™ Hand Cleaner
SDP

Catalog No. 19077516 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
2000 mL
Quantity:
4 Pk.
Packaging:
Refill
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity Packaging
19-077-516 2000 mL 4 Pk. Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-077-516 Supplier GOJO™ Supplier No. 726504
  • $49.75
    $37.65 / Each

    Save $12.10

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $173.00
    $130.65 / Case of 4 EA

    Save $42.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multi-purpose hand cleaner with natural pumice scrubbers.

  • Gel-style hand cleaner clings to hands and spreads easy
  • Contains natural pumice scrubbers
  • USDA BioPreferred™ Certified
  • Readily biodegradable
  • Contains a natural citrus solvent
  • Meets California VOC requirement for consumer products
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with every refill
  • Effective on oil, grease, paint, tar, and asphalt
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Cleanser
Color Green
Form Foam
Composition Laureth-7
For Use With (Application) For removing light to medium Industrial soils and dirt
Packaging Refill
pH Range 6 to 9
Quantity 4 Pk.
Size 2000 mL
Volume (English) 0.52 gal.
Volume (Metric) 2000 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.