Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ MULTI GREEN™ Hand Cleaner
SDP

Catalog No. p-3607732 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
800 mL
2000 mL
Quantity:
12 Pk.
4 Pk.
Packaging:
Dispenser Refills
Refill
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity Packaging
19-077-516 2000 mL 4 Pk. Refill
19-088-286 800 mL 12 Pk. Dispenser Refills
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-077-516 Supplier GOJO™ Supplier No. 726504
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Multi-purpose hand cleaner with natural pumice scrubbers.

  • Gel-style hand cleaner clings to hands and spreads easy
  • Contains natural pumice scrubbers
  • USDA BioPreferred™ Certified
  • Readily biodegradable
  • Contains a natural citrus solvent
  • Meets California VOC requirement for consumer products
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with every refill
  • Effective on oil, grease, paint, tar, and asphalt
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Cleanser
Color Green
Form Foam
Composition Laureth-7
For Use With (Application) For removing light to medium Industrial soils and dirt
Packaging Refill
pH Range 6 to 9
Quantity 4 Pk.
Size 2000 mL
Volume (English) 0.52 gal.
Volume (Metric) 2000 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.