Learn More
Description
- Cleans without harsh solvents
- Contains soothing emollients
- Rinses off or wipes clean with a towel
Specifications
Specifications
| Product Type | Hand Cleaner |
| Color | White |
| Composition | Petroleum Distillates (vapor), Mineral Oil (mist), Propylene Glycol, Oleic Acid |
| For Use With (Application) | For removing heavy grease, tar and oil |
| Packaging | Cartridge, plastic |
| Quantity | 6 Pk. |
| Volume (English) | 4.5 lb. |
| Volume (Metric) | 2 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
