Learn More
Enterprise Technology Solutions Gojo Soap,Antimicrobial,Foam,f/CS8 Touch-Free,1200ml,2/CT,CL
SDP
Catalog No. 5000830882
Encompass_Preferred
Antimicrobial foam soap fits in your PURELL CS8 Touch-Free Soap Dispenser. PURELL Healthy Soap 0.5 percent PCMX Antimicrobial Foam Handwash is effective in killing the most common bacteria found in healthcare environments. It helps reduce germs on skin that could cause disease. High-efficacy, high-quality, rich foam encourages frequent handwashing. It is designed for frequent use in healthcare environment among staff and patients. Dermatologist-tested, triclosan-free formulation is dye-free with pleasant floral fragrance. Foam soap rinses clean for easy gloving. It is compatible with nitrile, latex and vinyl gloves. With energy-on-the-refill technology, every refill has a built-in energy source, designed to last as long as the refill. Sanitary Sealed PET refill helps prevent contamination and is easily recycled.
Catalog No. 50-008-30882 Supplier Enterprise Technology Solutions Supplier No. GOJ787802
Antimicrobial foam soap fits in your PURELL CS8 Touch-Free Soap Dispenser. PURELL Healthy Soap 0.5 percent PCMX Antimicrobial Foam Handwash is effective in killing the most common bacteria found in healthcare environments. It helps reduce germs on skin that could cause disease. High-efficacy, high-quality, rich foam encourages frequent handwashing. It is designed for frequent use in healthcare environment among staff and patients. Dermatologist-tested, triclosan-free formulation is dye-free with pleasant floral fragrance. Foam soap rinses clean for easy gloving. It is compatible with nitrile, latex and vinyl gloves. With energy-on-the-refill technology, every refill has a built-in energy source, designed to last as long as the refill. Sanitary Sealed PET refill helps prevent contamination and is easily recycled.