Learn More
Enterprise Technology Solutions Gojo Soap,Foam,Antimicrobial,CS6 Dispenser,1200ml,2/CT,BN
SDP
Supplier:  Enterprise Technology Solutions GOJ658102
Encompass_Preferred
Designed for PURELL CS6 Touch-Free Soap Dispensers, this highly effective, antimicrobial handwash is tough on germs and gentle on hands. PURELL Healthcare Healthy Soap 2 percent CHG Antimicrobial Foam exceeds the FDA's Healthcare Personnel Handwash requirements and is proven as mild as GOJO's top-selling, nonantimicrobial soaps. Handwash is ideal for hospital-wide use or high-acuity areas. It is nonirritating, hypoallergenic and dermatologist-tested and contains chlorhexidine gluconate 2 percent - a broad-spectrum antimicrobial agent that kills pathogenic, gram-positive and gram-negative bacteria. Mild, dye-free and fragrance-free formulation with rich lather helps maintain the skin's natural oils and moisture. Sanitary Sealed PET refill is easily recycled.
Catalog No. 50-008-10717