Learn More
Enterprise Technology Solutions Gojo Soap Refill,Antibacterial,Foam,f/PROVONFMX-12,1250ml,AR
SDP
Catalog No. 5000831582
Encompass_Preferred
High-performance, antimicrobial foam soap contains added moisturizers for general handwashing. It is made with 0.3 percent chloroxylenol (PCMX), a common antimicrobial compound to help control bacteria. Foaming hand soap is pre-lathered for convenience and offers an appealing translucent amber color with light fruity fragrance. 1250 ml. refill is designed for use with PROVON FMX-12 Dispensers. Sanitary Sealed refill helps lock out germs. Each refill includes a fresh dispensing valve.
Catalog No. 50-008-31582 Supplier Enterprise Technology Solutions Supplier No. GOJ518604
High-performance, antimicrobial foam soap contains added moisturizers for general handwashing. It is made with 0.3 percent chloroxylenol (PCMX), a common antimicrobial compound to help control bacteria. Foaming hand soap is pre-lathered for convenience and offers an appealing translucent amber color with light fruity fragrance. 1250 ml. refill is designed for use with PROVON FMX-12 Dispensers. Sanitary Sealed refill helps lock out germs. Each refill includes a fresh dispensing valve.