Learn More
Enterprise Technology Solutions Gojo Soap Refill, Foodservice, f/ES6 Disp, 1200ml, 2/CT, BE
SDP
Catalog No. 5000831148
Encompass_Preferred
Effective antimicrobial, foam hand soap for your PURELL ES6 Touch-Free Soap Dispenser is specially formulated for foodservice environments to help remove food soils and odors. Luxurious foam formulation kills 99.99 of percent germs including the most common foodborne bacterial pathogens. Lightly fragranced foam soap for frequent handwashing is proven to be as mild as PURELL's selling non-antimicrobial soap. Dye-free, fragrance-free formula is enriched with a moisturizer, natural extract and skin conditioner. It contains no phthalates, parabens or triclosan. PURELL HEALTHY SOAP 0.5 percent BAK Antimicrobial Foam meets USDA biobased requirements for handwash. AT-A-GLANCE refill design lets you monitor soap levels with just one look. Sanitary Sealed PET refill is easily recycled.
Catalog No. 50-008-31148 Supplier Enterprise Technology Solutions Supplier No. GOJ648002
Effective antimicrobial, foam hand soap for your PURELL ES6 Touch-Free Soap Dispenser is specially formulated for foodservice environments to help remove food soils and odors. Luxurious foam formulation kills 99.99 of percent germs including the most common foodborne bacterial pathogens. Lightly fragranced foam soap for frequent handwashing is proven to be as mild as PURELL's selling non-antimicrobial soap. Dye-free, fragrance-free formula is enriched with a moisturizer, natural extract and skin conditioner. It contains no phthalates, parabens or triclosan. PURELL HEALTHY SOAP 0.5 percent BAK Antimicrobial Foam meets USDA biobased requirements for handwash. AT-A-GLANCE refill design lets you monitor soap levels with just one look. Sanitary Sealed PET refill is easily recycled.