Learn More
Enterprise Technology Solutions Gojo Spill Kit,f/Body Fluids,w/User Protection,2 Single-Uses/Kit
SDP
Catalog No. 5000828071
Encompass_Preferred
Simplify your response process to body fluid spills with PURELL Body Fluid Spill Kits. Each kit features PURELL Foodservice Surface Sanitizer, proven on a variety of hard and soft surfaces, including food-contact surfaces with no rinsing required. Quickly kill 99.9 percent of common germs that may cause illness with the surface sanitizer. One-step formula complies with FDA Food Code and OSHA. The included head-to-toe personal protection and cleanup products ensure you can safely remove vomit, blood, feces, other body fluids and broken glass. Each fully disposable kit helps avoid unnecessary biohazardous waste pickup fees and reduces cleanup time.
Catalog No. 50-008-28071 Supplier Enterprise Technology Solutions Supplier No. GOJ384108CLMS
Simplify your response process to body fluid spills with PURELL Body Fluid Spill Kits. Each kit features PURELL Foodservice Surface Sanitizer, proven on a variety of hard and soft surfaces, including food-contact surfaces with no rinsing required. Quickly kill 99.9 percent of common germs that may cause illness with the surface sanitizer. One-step formula complies with FDA Food Code and OSHA. The included head-to-toe personal protection and cleanup products ensure you can safely remove vomit, blood, feces, other body fluids and broken glass. Each fully disposable kit helps avoid unnecessary biohazardous waste pickup fees and reduces cleanup time.