Learn More
Medchemexpress LLC Gol-ntr | 2968461-58-5 | 99.4% | 467.40 | C24H16F3N3O4 | 10 MG
SDP
Catalog No. 5000286731
Encompass_Preferred
Gol-NTR is a Golgi-targetable fluorescent probe activated by nitroreductase (NTR), designed for selective and sensitive imaging of NTR activity in cells and in vivo. The probe is intended for fluorescent labeling and monitoring of disease models where NTR activity is relevant, including acute lung injury research.
  • Golgi-targetable probe for subcellular imaging
  • NTR-activated fluorescence for selective detection
  • Low detection limit of 54.8 ng/mL for high sensitivity
  • Suitable for fluorescent labeling in A549 cells and in vivo imaging
  • High reported purity (99.4%) for research applications
  • Solid, white to off-white form with recommended sealed storage
Catalog No. 50-002-86731 Supplier Medchemexpress LLC Supplier No. HY15153710MG
Gol-NTR is a Golgi-targetable fluorescent probe activated by nitroreductase (NTR), designed for selective and sensitive imaging of NTR activity in cells and in vivo. The probe is intended for fluorescent labeling and monitoring of disease models where NTR activity is relevant, including acute lung injury research.
  • Golgi-targetable probe for subcellular imaging
  • NTR-activated fluorescence for selective detection
  • Low detection limit of 54.8 ng/mL for high sensitivity
  • Suitable for fluorescent labeling in A549 cells and in vivo imaging
  • High reported purity (99.4%) for research applications
  • Solid, white to off-white form with recommended sealed storage